Learn More
Invitrogen™ PMEPA1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120147
Description
Positive test controls include: SKOV3, 22Rv1, 293T, Mouse brain, Rat brain, Rat heart. The target is usually found in the following locations: Early endosome membrane, Golgi apparatus membrane, Single-pass membrane protein. Immunogen sequence: PSSNSGISAT CYGSGGRMEG PPPTYSEVIG HYPGSSFQHQ QSSGPPSLLE GTRLHHTHIA PLESAAIWSK EKDKQKGHPL.
This gene encodes a transmembrane protein that contains a Smad interacting motif (SIM). Expression of this gene is induced by androgens and transforming growth factor beta, and the encoded protein suppresses the androgen receptor and transforming growth factor beta signaling pathways though interactions with Smad proteins. Overexpression of this gene may play a role in multiple types of cancer. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Specifications
PMEPA1 | |
Polyclonal | |
Unconjugated | |
PMEPA1 | |
2210418I02Rik; AW455466; Erg1.2; N4wbp4; Nedd4 WW binding protein 4; NEDD4 WW domain-binding protein 4; PMEPA1; Prostate transmembrane protein androgen induced 1; prostate transmembrane protein, androgen induced 1; protein TMEPAI; Solid tumor-associated 1 protein; STAG1; Tmepai; transmembrane prostate androgen-induced protein; transmembrane, prostate androgen induced RNA | |
Rabbit | |
Affinity Chromatography | |
RUO | |
311676, 56937, 65112 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
3.33 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q969W9, Q9D7R2 | |
PMEPA1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 173-252 of human PMEPA1 (NP_954638.1). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.