Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PMF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567206
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567206 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567206 Supplier Thermo Scientific Supplier No. PA567206
Add to Cart
Add to Cart

PMF1 Polyclonal Antibody

PMF1 Polyclonal Antibody for ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen PMF1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PMF1
Gene Accession No. Q6P1K2
Gene Alias 2600009M07Rik; AL033286; AW060657; Est1p-like protein B (EST1B); Pmf1; PMF-1; polyamine modulated factor 1; polyamine-modulated factor 1; RP11-54H19.4; Unknown (protein for MGC:134238)
Gene Symbols PMF1
Host Species Rabbit
Immunogen Recombinant Human PMF1. Recombinant protein control fragment (Product #RP-105548).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11243
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.