Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PMFBP1 Monoclonal Antibody (4G9)
GREENER_CHOICE

Catalog No. PIMA524491
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524491 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524491 Supplier Invitrogen™ Supplier No. MA524491
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: FHTEELQTSY YSLRQYQSIL EKQTSDLVLL HHHCKLKEDE VILYEEEMGN HNENTGEKLH LAQEQLALAG DKIASLERSL NLYRDKYQSS LSNIELLEC.

May play a role in sperm morphology especially the sperm tail and consequently affect fertility. May also be involved in the general organization of cellular cytoskeleton.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PMFBP1
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4G9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene PMFBP1
Gene Accession No. Q8TBY8
Gene Alias 1700016D22Rik; F77; LOW QUALITY PROTEIN: polyamine-modulated factor 1-binding protein 1; ODF3; outer dense fiber ODF3; outer dense fiber protein 3; PMF-1 binding protein; PMF-1-binding protein; PMFBP1; polyamine modulated factor 1 binding protein 1; polyamine-modulated factor 1-binding protein 1; sperm tail associated protein; sperm tail-associated protein; Stap
Gene Symbols PMFBP1
Host Species Mouse
Immunogen PMFBP1 (NP_112583, 99 a.a. ~ 197 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83449
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.