Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PMFBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566035
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566035 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566035 Supplier Invitrogen™ Supplier No. PA566035
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LYRDKYQSSLS NIELLECQVK MLQGELGGIM GQEPENKGDH SKVRIYTSPC MIQEHQETQK RLSEVWQKVS QQDDLIQELR NKLACSNALV LEREKALIKL QADFASCTAT HRYPPSSSEE CEDIKKILKH LQEQKD Highest antigen sequence identity to the following orthologs - mouse 87%, rat 86%.

May play a role in sperm morphology especially the sperm tail and consequently affect fertility. May also be involved in the general organization of cellular cytoskeleton.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PMFBP1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PMFBP1
Gene Accession No. Q8TBY8
Gene Alias 1700016D22Rik; F77; LOW QUALITY PROTEIN: polyamine-modulated factor 1-binding protein 1; ODF3; outer dense fiber ODF3; outer dense fiber protein 3; PMF-1 binding protein; PMF-1-binding protein; PMFBP1; polyamine modulated factor 1 binding protein 1; polyamine-modulated factor 1-binding protein 1; sperm tail associated protein; sperm tail-associated protein; Stap
Gene Symbols PMFBP1
Host Species Rabbit
Immunogen Recombinant Human PMFBP1. Recombinant protein control fragment (Product #RP-106723).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83449
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.