Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ PML Isoform II, Mouse, Unlabeled, Clone: 1A8.1,

Catalog No. MABE175MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABE175MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE175MI Supplier MilliporeSigma™ Supplier No. MABE175
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Protein PML (UniProt P29590; also known as Promyelocytic leukemia protein, RING finger protein 71, Tripartite motif-containing protein 19) is encoded by the PML (also known as MYL, PP8675, RNF71, TRIM19) gene (Gene ID 5371) in human. The human PML gene/protein adopted its name when it was originally discovered as an RARα fusion protein caused by chromosomal translocation in acute promyelocytic leukemia (APL) patients. In normal cells, the PML protein interacts with multiple protein partners in the nucleus to form PML nuclear bodies (PML-NBs) that mediate a variety of cellular functions including senescence, apoptosis, protein degradation, telemere stasbility, and antiviral defense. At least 12 PML isoforms are generated by alternative splicings, including six nuclear isoforms (PML-I to PML-VI) and the cytoplasmic isoform PML-VII (a.k.a. PML-VIIB), other spliced isoforms wihtout exon 6-coding sequence (a.a. 467-552) are likewise expected to be cytoplasmic due to the abscence of nuclear localization signal (NLS) encoded by exon 6. All isoforms share the same N-terminal region (a.a. 1-419) containing the RBCC/tripartite motif and may have related functions due to their common functional RBCC/TRIM domain, while increasing evidences also suggest that each PML isoform possesses distinct functions mediated by its specific C-terminal sequence. Isoform PML-1 (a.k.a. PML-I, TRIM19alpha; UniProt P29590-1) and PML-2 (a.k.a. PML-II or TRIM19kappa; UniProt P29590-8) share the same N-terminal 570-a.a. sequence (encoded by exons 1 to 7a), however PML-1 derives its remaining C-terminal sequence (a.a. 571-882) from exons 8a and 9, while PML-II derives its (a.a. 571-829) from exon 7b with a different reading frame as that of PML-V.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PML Isoform II
Applications Western Blot
Classification Monoclonal
Clone 1A8.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene PML, MYL, PP8675, RNF71, TRIM19
Gene Accession No. P29590-8
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to a C-terminal sequence of human PML Isoform II.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_150242
Test Specificity Clone 1A8.1 was raised against an C-terminal region sequence unique to PML isoform II (PML-2), but not present in the other 11 PML spliced isoforms reported by UniProt (P29590).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.