Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PML Polyclonal Antibody, Bioss
SDP

Catalog No. 502280687
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-228-0687 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-228-0687 Supplier Bioss Supplier No. BS53134R
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PML Polyclonal antibody specifically detects PML in Human samples. It is validated for ELISA, Western Blot

Specifications

Antigen PML
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.755 mg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene PML
Gene Accession No. P29590
Gene Alias 1200009E24Rik; AI661194; MYL; Pml; PML/RARA fusion; pml1; PP8675; probable transcription factor PML; promyelocytic leukemia; promyelocytic leukemia protein; promyelocytic leukemia, inducer of; protein PML; RGD1562602; RING finger protein 71; RNF71; TRIM19; tripartite motif protein TRIM19; tripartite motif-containing protein 19
Host Species Rabbit
Immunogen Polyclonal antibody raised in rabbit against human PML (Promyelocytic leukemia), using 3 different KLH-conjugated synthetic peptides
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5371
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.