Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PMM1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556547
Description
Immunogen sequence: MAVTAQAARR KERVLCLFDV DGTLTPARQK IDPEVAAFLQ KLRSRVQIGV VGGSDY Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 88%.
PMM1, also named as PMMH22, belongs to the eukaryotic PMM family. It catalyzes the second step in the conversion of fructose-6P to GDP-mannose is and is involved in the synthesis of the GDP-mannose and dolichol-phosphate-mannose required for a number of critical mannosyl transfer reactions. In addition, PMM1 may be responsible for the degradation of glucose-1, 6-bisphosphate in ischemic brain.
Specifications
PMM1 | |
Polyclonal | |
Unconjugated | |
Pmm1 | |
brain glucose-1,6-bisphosphatase; C77612; Phosphomannomutase 1; PMM 1; Pmm1; PMMH22; PMMH-22; Sec53; Secp53 homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29858, 300089, 5372 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O35621, Q92871 | |
Pmm1 | |
Recombinant protein corresponding to Human PMM1. Recombinant protein control fragment (Product #RP-95363). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction