Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PMM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5139929
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5139929 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5139929 Supplier Invitrogen™ Supplier No. PA5139929
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: YCLDSLDQDS FDTIHFFGNE TSPGGNDFEI FADPRTVGHS VVSPQDTVQR CREIFFPETA HEA Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 94%.

PMM1, also named as PMMH22, belongs to the eukaryotic PMM family. It catalyzes the second step in the conversion of fructose-6P to GDP-mannose is and is involved in the synthesis of the GDP-mannose and dolichol-phosphate-mannose required for a number of critical mannosyl transfer reactions. In addition, PMM1 may be responsible for the degradation of glucose-1, 6-bisphosphate in ischemic brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PMM1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Pmm1
Gene Accession No. Q92871
Gene Alias brain glucose-1,6-bisphosphatase; C77612; Phosphomannomutase 1; PMM 1; Pmm1; PMMH22; PMMH-22; Sec53; Secp53 homolog
Gene Symbols Pmm1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PMM1. Recombinant protein control fragment (Product #RP-109444).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5372
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.