Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PMM2/Phosphomannomutase 2 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300351PCP
Description
PMM2/Phosphomannomutase 2 Polyclonal antibody specifically detects PMM2/Phosphomannomutase 2 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
PMM2/Phosphomannomutase 2 | |
Polyclonal | |
Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry-Paraffin | |
CDG1, CDG1A, CDGS, EC 5.4.2.8, phosphomannomutase 2, PMM 2 | |
Produced in rabbits immunized with purified, recombinant Human PMM2/Phosphomannomutase 2 (Accession#: O15305; Met1-Ser246) | |
0.1 mL | |
Lipid and Metabolism | |
5373 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction