Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PMM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566612
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566612 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566612 Supplier Invitrogen™ Supplier No. PA566612
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FEKVQEQLGND VVEKYDYVFP ENGLVAYKDG KLLCRQNIQS Highest antigen sequence identity to the following orthologs - mouse 93%, rat 93%.

The protein encoded by this gene catalyzes the isomerization of mannose 6-phosphate to mannose 1-phosphate, which is a precursor to GDP-mannose necessary for the synthesis of dolichol-P-oligosaccharides. Mutations in this gene have been shown to cause defects in glycoprotein biosynthesis, which manifests as carbohydrate-deficient glycoprotein syndrome type I.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PMM2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PMM2
Gene Accession No. O15305
Gene Alias AI585868; C86848; CDG1; CDG1a; CDGS; mannose-6-phosphate isomerase; phosphomannomutase 2; phosphomannose isomerase 1; PMI; PMI1; PMM 2; Pmm2
Gene Symbols PMM2
Host Species Rabbit
Immunogen Recombinant Human PMM2. Recombinant protein control fragment (Product #RP-105266).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5373
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.