Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PMP22 Antibody (Hu1), DyLight 550

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11059086R
Description
PMP22 Monoclonal specifically detects PMP22 in Human, Primate, Bovine (Negative), Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PMP22 | |
Monoclonal | |
DyLight 550 | |
Q01453 | |
PMP22 | |
Human PMP22 cDNA boosted with a 13-amino acid peptide of the 2nd extracellular domain (a.a. 120-133) conjugated to murine IgG. [Swiss-Prot# Q01453] | |
0.1 mL | |
Cellular Markers, Glia Markers, Membrane Trafficking and Chaperones, Neuroscience, Oligodendrocyte Cell Markers | |
5376 | |
Human, Primate, Bovine (Negative), Mouse (Negative), Rat (Negative) | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Hu1 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
CMT1A, CMT1E, GAS3, GAS-3DSS, growth arrest-specific 3, Growth arrest-specific protein 3, HMSNIA, HNPP, MGC20769, peripheral myelin protein 22, PMP-22, Sp110 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human and Rhesus monkey. Does not react with mouse, rat, or bovine nerve protein. | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction