Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PMP22 Antibody (Hu1), DyLight 755

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11059086IR
Description
PMP22 Monoclonal specifically detects PMP22 in Human, Primate, Bovine (Negative), Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PMP22 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PMP22 | |
Human PMP22 cDNA boosted with a 13-amino acid peptide of the 2nd extracellular domain (a.a. 120-133) conjugated to murine IgG. [Swiss-Prot# Q01453] | |
0.1 mL | |
Primary | |
Human, Primate, Bovine (Negative), Mouse (Negative), Rat (Negative) | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Hu1 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
CMT1A, CMT1E, GAS3, GAS-3DSS, growth arrest-specific 3, Growth arrest-specific protein 3, HMSNIA, HNPP, MGC20769, peripheral myelin protein 22, PMP-22, Sp110 | |
Mouse | |
Protein G purified | |
Cellular Markers, Glia Markers, Membrane Trafficking and Chaperones, Neuroscience, Oligodendrocyte Cell Markers | |
5376 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction