Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PMP22 Mouse anti-Human, Clone: OTI5D1, TrueMAB™, Origene
SDP

Catalog No. 502243733
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-224-3733 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-224-3733 Supplier OriGene Supplier No. TA808964S
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PMP22 Monoclonal antibody specifically detects PMP22 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen PMP22
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI5D1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PMP22
Gene Accession No. Q01453
Gene Alias 22kDa; Cd25; CMT1A; CMT1E; DSS; GAS3; GAS-3; Growth arrest-specific protein 3; HMSNIA; HNPP; peripheral myelin; peripheral myelin protein 22; peripheral myelin protein 22 kDa; PMP22; PMP-22; Protein CD25; SAG; schwann cell membrane glycoprotein; Sp110; SR13 myelin protein; Tr; trembler
Host Species Mouse
Immunogen Full length human recombinant protein of human PMP22 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5376
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.