Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PMP22 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599626
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599626 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599626 Supplier Thermo Scientific Supplier No. PA599626
Only null left
Add to Cart
Add to Cart

PMP22 Polyclonal Antibody

PMP22 Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen PMP22
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PMP22
Gene Accession No. P16646, P25094, Q01453
Gene Alias 22kDa; Cd25; CMT1A; CMT1E; DSS; GAS3; GAS-3; Growth arrest-specific protein 3; HMSNIA; HNPP; peripheral myelin; peripheral myelin protein 22; peripheral myelin protein 22 kDa; PMP22; PMP-22; Protein CD25; SAG; schwann cell membrane glycoprotein; Sp110; SR13 myelin protein; Tr; trembler
Gene Symbols PMP22
Host Species Rabbit
Immunogen A synthesized peptide derived from human PMP22(Accession Q01453), corresponding to amino acid residues P122-E160.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18858, 24660, 5376
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.