Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PMP22 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980165
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0165 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0165 Supplier Bioss Supplier No. BS0235R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peripheral Myelin Protein 22 (PMP22) is a 160 amino acid glycoprotein which is a member of the claudin family and is thought to play an important role in the structure of peripheral nerve myelin. Irregularities in its gene results in hereditary demyelinating neuropathies. PMP22 protein is present in the compact myelin of adult human peripheral nerve and its distribution mimics that of Myelin Protein zero (P0).

Specifications

Antigen PMP22
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PMP22
Gene Accession No. Q01453
Gene Alias 22kDa; Cd25; CMT1A; CMT1E; DSS; Gas3; Gas-3; Growth arrest-specific protein 3; HMSNIA; HNPP; Peripheral myelin protein 22; peripheral myelin protein 22 kDa; Pmp22; Pmp-22; Protein CD25; SAG; Schwann cell membrane glycoprotein; Sp110; SR13 myelin protein; Tr; trembler
Gene Symbols PMP22
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PMP-22.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5376
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.