Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PMP22 Recombinant Rabbit Monoclonal Antibody (JM52-30)

Rabbit Recombinant Monoclonal Antibody

Supplier:  Invitrogen™ MA532726

Catalog No. PIMA532726


Only null left
Add to Cart

Description

Description

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Peripheral Myelin Protein 22 (PMP22) is a 160 amino acid glycoprotein which is a member of the claudin family and is thought to play an important role in the structure of peripheral nerve myelin. Irregularities in its gene results in hereditary demyelinating neuropathies. PMP22 protein is present in the compact myelin of adult human peripheral nerve and its distribution mimics that of Myelin Protein zero (P0).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PMP22
Recombinant Monoclonal
1 mg/mL
TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
P25094, Q01453
PMP22
Synthetic peptide within Human PMP22 aa 1-50.
100 μL
Primary
Human, Rat
Antibody
IgG
Western Blot, Immunocytochemistry, Immunohistochemistry, Western Blot, Western Blot
JM52-30
Unconjugated
PMP22
22kDa; Cd25; CMT1A; CMT1E; DSS; GAS3; GAS-3; Growth arrest-specific protein 3; HMSNIA; HNPP; peripheral myelin; peripheral myelin protein 22; peripheral myelin protein 22 kDa; PMP22; PMP-22; Protein CD25; SAG; schwann cell membrane glycoprotein; Sp110; SR13 myelin protein; Tr; trembler
Rabbit
Protein A
RUO
24660, 5376
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.