Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PMP70 Polyclonal Antibody, Alexa Fluor™ 647
GREENER_CHOICE

Catalog No. PIPA1650AY
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA1650AY 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA1650AY Supplier Invitrogen™ Supplier No. PA1650A647
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

PA1650A647 immunizing peptide corresponds to amino acid residues 644-659 from rat PMP70. This sequence is completely conserved in mouse PMP70 and differs from human PMP70 by a single amino acid substitution. The immunizing peptide (Cat. # PEP-038) is available for use in neutralization and control experiments.

Peroxisomes are single membrane organelles which are involved with many important biochemical pathways and are found in almost all eukaryotic cells. Various toxins including, phenols, alcohols, and aldehydes are targeted for modification by peroxisomes. Long chain fatty acid metabolism via beta-oxidation is also mediated by this organelle. Loss of peroxisomal functions may result in diseases such as Zellweger syndrome, hyperpipecolic acidemia, neonatal adrenoleukodystrophy, and infantile Refsum' disease. Peroxisomal membrane protein 70 (PMP70) is an abundant, integral membrane protein of the peroxisome. This protein is induced by treatment with hypolipidemic agents in parallel with peroxisome proliferation and stimulation of the peroxisomal beta-oxidation enzymes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PMP70
Applications Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Alexa Fluor 647
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4
Gene ABCD3
Gene Accession No. P16970, P28288, P55096
Gene Alias 68 kDa peroxisomal membrane protein; 70 kDa peroxisomal membrane protein; 70-kDa peroxisomal membrane protein; ABC43; ABCD3; AI313901; ATP binding cassette subfamily D member 3; ATP-binding cassette sub-family D member 3; ATP-binding cassette, subfamily D (ALD), member 3; ATP-binding cassette, sub-family D (ALD), member 3; AU018866; AW146054; CBAS5; dJ824O18.1 (ATP-binding cassette, sub-family D (ALD), member 3 (PMP70, PXMP1)); Peroxisomal membrane protein 1; peroxisomal membrane protein 1 (70kD, Zellweger syndrome); peroxisomal membrane protein, 70 kDa; Peroxisomal membrane protein-1 (70kD); PMP68; Pmp70; Pxmp1; ZWS2
Gene Symbols ABCD3
Host Species Rabbit
Immunogen Synthetic Peptide: C N(644) Y E F K K I T E D T V E F G S(659).
Purification Method Antigen affinity chromatography
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19299, 25270, 5825
Target Species Human, Mouse, Rat
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.