Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PMP70 Polyclonal Antibody, Biotin
GREENER_CHOICE

Catalog No. PIPA5142389
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142389 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142389 Supplier Invitrogen™ Supplier No. PA5142389
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Additional Information: This antibody is expected to recognize one isoform (NP_002849.1) only. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.

Peroxisomes are single membrane organelles which are involved with many important biochemical pathways and are found in almost all eukaryotic cells. Various toxins including, phenols, alcohols, and aldehydes are targeted for modification by peroxisomes. Long chain fatty acid metabolism via beta-oxidation is also mediated by this organelle. Loss of peroxisomal functions may result in diseases such as Zellweger syndrome, hyperpipecolic acidemia, neonatal adrenoleukodystrophy, and infantile Refsum disease. Peroxisomal membrane protein 70 (PMP70) is an abundant, integral membrane protein of the peroxisome. This protein is induced by treatment with hypolipidemic agents in parallel with peroxisome proliferation and stimulation of the peroxisomal beta-oxidation enzymes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PMP70
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene ABCD3
Gene Accession No. P28288
Gene Alias 68 kDa peroxisomal membrane protein; 70 kDa peroxisomal membrane protein; 70-kDa peroxisomal membrane protein; ABC43; ABCD3; AI313901; ATP binding cassette subfamily D member 3; ATP-binding cassette sub-family D member 3; ATP-binding cassette, subfamily D (ALD), member 3; ATP-binding cassette, sub-family D (ALD), member 3; AU018866; AW146054; CBAS5; dJ824O18.1 (ATP-binding cassette, sub-family D (ALD), member 3 (PMP70, PXMP1)); Peroxisomal membrane protein 1; peroxisomal membrane protein 1 (70kD, Zellweger syndrome); peroxisomal membrane protein, 70 kDa; Peroxisomal membrane protein-1 (70kD); PMP68; Pmp70; Pxmp1; ZWS2
Gene Symbols ABCD3
Host Species Goat
Immunogen Peptide with sequence C-PDGREDQKRKGISD.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5825
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.