Learn More
Invitrogen™ PMP70 Recombinant Rabbit Monoclonal Antibody (3V4K3)

Description
Positive Samples: HepG2, A-549, Mouse liver, Mouse kidney, Rat liver Immunogen sequence: GWDSVQDWMD VLSGGEKQRM AMARLFYHKP QFAILDECTS AVSVDVEGYI YSHCRKVGIT LFTVSHRKSL WKHHEYYLHM DGRGNYEFKQ ITEDTVEFGS.
Specifications
Specifications
| Antigen | PMP70 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 3V4K3 |
| Concentration | 0.8 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | ABCD3 |
| Gene Accession No. | P16970, P28288, P55096 |
| Gene Alias | 68 kDa peroxisomal membrane protein; 70 kDa peroxisomal membrane protein; 70-kDa peroxisomal membrane protein; ABC43; ABCD3; AI313901; ATP binding cassette subfamily D member 3; ATP-binding cassette sub-family D member 3; ATP-binding cassette, subfamily D (ALD), member 3; ATP-binding cassette, sub-family D (ALD), member 3; AU018866; AW146054; CBAS5; dJ824O18.1 (ATP-binding cassette, sub-family D (ALD), member 3 (PMP70, PXMP1)); Peroxisomal membrane protein 1; peroxisomal membrane protein 1 (70kD, Zellweger syndrome); peroxisomal membrane protein, 70 kDa; Peroxisomal membrane protein-1 (70kD); PMP68; Pmp70; Pxmp1; ZWS2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.