Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PMPCA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554481
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554481 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554481 Supplier Invitrogen™ Supplier No. PA554481
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VEHEHLVDCA RKYLLGVQPA WGSAEAVDID RSVAQYTGGI AKLERDMSNV SLGPTPIPEL THIMVGLESC SFLEEDFIPF AVLNMMMGGG GSFSAGGPGK GMFSR Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 92%.

Substrate recognition and binding subunit of the essential mitochondrial processing protease (MPP), which is required for maturation of the majority of mitochondrial precursor proteins. Most MPP cleavage sites follow an arginine at position -2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PMPCA
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PMPCA
Gene Accession No. Q10713
Gene Alias 1200002L24Rik; 4933435E07Rik; alpha-MPP; inactive zinc metalloprotease alpha; inositol polyphosphate-5-phosphatase, 72 kD; INPP5E; KIAA0123; mitochondrial matrix processing protease, alpha subunit; mitochondrial processing peptidase alpha subunit, mitochondrial; mitochondrial-processing peptidase subunit alpha; MPPA; P-55; peptidase (mitochondrial processing) alpha; peptidase, mitochondrial processing alpha subunit; peptidase, mitochondrial processing subunit alpha; PMPCA; SCAR2; Unknown (protein for MGC:139568)
Gene Symbols PMPCA
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PMPCA. Recombinant protein control fragment (Product #RP-93048).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23203
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.