Learn More
PMPCA Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | PMPCA |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-0648
![]() |
Bethyl Laboratories, Inc
A305520AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine and Orangutan
PMPCA cleaves presequences (transit peptides) from mitochondrial protein precursors.Specifications
PMPCA | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Alpha-MPP, inositol polyphosphate-5-phosphatase, 72 kD, INPP5E, KIAA0123, mitochondrial matrix processing protease, alpha subunit, P-55, peptidase (mitochondrial processing) alpha, SCAR2 | |
PMPCA | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q10713 | |
23203 | |
Between 25 and 75 | |
Primary | |
4° C | |
PMPCA |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.