Learn More
PMPCA Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305556AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat and Orangutan
PMPCA cleaves presequences (transit peptides) from mitochondrial protein precursors.Specifications
PMPCA | |
Polyclonal | |
Unconjugated | |
PMPCA | |
Alpha-MPP, inositol polyphosphate-5-phosphatase, 72 kD, INPP5E, KIAA0123, mitochondrial matrix processing protease, alpha subunit, P-55, peptidase (mitochondrial processing) alpha, SCAR2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23203 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q10713 | |
PMPCA | |
Between 475 and 525 | |
10 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.