Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PMPCB Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559246
Description
Immunogen sequence: WGFSESLLIR GAAGRSLYFG ENRLRSTQAA TQVVLNVPET RVTCLESGLR VASEDSGLST CTVGLWIDAG Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 77%.
PMPCB is a member of the peptidase M16 family and encodes a protein with a zinc-binding motif. This protein is located in the mitochondrial matrix and catalyzes the cleavage of the leader peptides of precursor proteins newly imported into the mitochondria, though it only functions as part of a heterodimeric complex.
Specifications
PMPCB | |
Polyclonal | |
Unconjugated | |
PMPCB | |
3110004O18Rik; Beta-MPP; mitochondrial processing peptidase beta subunit; mitochondrial-processing peptidase subunit beta; MPP11; MPPB; MPPP52; P-52; peptidase (mitochondrial processing) beta; peptidase, mitochondrial processing beta subunit; PMPCB | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9512 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75439 | |
PMPCB | |
Recombinant protein corresponding to Human PMPCB. Recombinant protein control fragment (Product #RP-98371). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction