Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PMPCB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116547
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116547 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116547 Supplier Invitrogen™ Supplier No. PA5116547
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: LO2, HT-1080, Mouse kidney, Rat liver Immunogen sequence: GTKKRSQLDL ELEIENMGAH LNAYTSREQT VYYAKAFSKD LPRAVEILAD IIQNSTLGEA EIERERGVIL REMQEVETNL QEVVFDYLHA TAYQNTALGR TILGPTENIK SISRKDLVDY ITTHYKGPRI VLAAAGGVSH DELLDLAKFH FGDSLCTHKG EIPALPPCKF TGSEIRVRDD KMPLAHLAIA VEAVGWAHPD TICLMVANTL IGNWDRSFGG GMNLSSKLAQ LTCHGNLCHS FQSFNTSYTD TGLWGLYMVC ESSTVADMLH V.

PMPCB is a member of the peptidase M16 family and encodes a protein with a zinc-binding motif. This protein is located in the mitochondrial matrix and catalyzes the cleavage of the leader peptides of precursor proteins newly imported into the mitochondria, though it only functions as part of a heterodimeric complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PMPCB
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.46 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PMPCB
Gene Accession No. O75439, Q03346, Q9CXT8
Gene Alias 3110004O18Rik; Beta-MPP; mitochondrial processing peptidase beta subunit; mitochondrial-processing peptidase subunit beta; MPP11; MPPB; MPPP52; P-52; peptidase (mitochondrial processing) beta; peptidase, mitochondrial processing beta subunit; PMPCB
Gene Symbols PMPCB
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 110-380 of human PMPCB (NP_0042702).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64198, 73078, 9512
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.