Learn More
PMS1 Monoclonal Antibody (OTI5E10)

Description
PMS1 Monoclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | PMS1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI5E10 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | PMS1 |
| Gene Accession No. | P54277 |
| Gene Alias | DNA mismatch repair protein PMS1; HNPCC3; hPMS1; human homolog of yeast mutL; mismatch repair gene PMSL1; MLH2; PMS1; PMS1 homolog 1, mismatch repair system component; PMS1 postmeiotic segregation increased 1; PMS1 protein homolog 1; PMSL1; postmeiotic segregation increased 1 (S. cerevisiae); rhabdomyosarcoma antigen MU-RMS-40.10B; rhabdomyosarcoma antigen MU-RMS-40.10E |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.