Learn More
PMS1 Mouse anti-Human, Clone: OTI5E10, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF810414

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a protein belonging to the DNA mismatch repair mutL/hexB family. This protein is thought to be involved in the repair of DNA mismatches, and it can form heterodimers with MLH1, a known DNA mismatch repair protein. Mutations in this gene cause hereditary nonpolyposis colorectal cancer type 3 (HNPCC3) either alone or in combination with mutations in other genes involved in the HNPCC phenotype, which is also known as Lynch syndrome.Specifications
PMS1 | |
Monoclonal | |
Unconjugated | |
PMS1 | |
HNPCC3, MLH2, PMSL1, hPMS1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
5378 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI5E10 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P54277 | |
PMS1 | |
Human recombit protein fragment corresponding to amino acids 323-423 of human PMS1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.