Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PMS1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986240
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6240 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6240 Supplier Bioss Supplier No. BS4965R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a protein belonging to the DNA mismatch repair mutL/hexB family. This protein is thought to be involved in the repair of DNA mismatches, and it can form heterodimers with MLH1, a known DNA mismatch repair protein. Mutations in this gene cause hereditary nonpolyposis colorectal cancer type 3 (HNPCC3) either alone or in combination with mutations in other genes involved in the HNPCC phenotype, which is also known as Lynch syndrome.

Specifications

Antigen PMS1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PMS1
Gene Accession No. P54277
Gene Alias DNA mismatch repair protein PMS1; HNPCC3; hPMS1; human homolog of yeast mutL; mismatch repair gene PMSL1; MLH2; PMS1; PMS1 homolog 1, mismatch repair system component; PMS1 postmeiotic segregation increased 1; PMS1 protein homolog 1; PMSL1; postmeiotic segregation increased 1 (S. cerevisiae); rhabdomyosarcoma antigen MU-RMS-40.10B; rhabdomyosarcoma antigen MU-RMS-40.10E
Gene Symbols PMS1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PMS1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5378
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.