Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PMS2 Antibody (OTI2G5), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273481FR
Description
PMS2 Monoclonal specifically detects PMS2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
PMS2 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
PMS2 | |
Full length recombinant protein of human PMS2 (NP_000526) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG2b |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI2G5 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
DNA mismatch repair protein PMS2, EC 3.1, H_DJ0042M02.9, HNPCC4postmeiotic segregation increased (S. cerevisiae) 2, mismatch repair endonuclease PMS2, PMS1 protein homolog 2, PMS2 postmeiotic segregation increased 2 (S. cerevisiae), PMSL2PMS2CL | |
Mouse | |
Affinity Purified | |
Cancer, Cell Cycle and Replication, DNA Repair, Mismatch Repair | |
5395.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction