Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PMS2 Antibody (OTI2G5), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273481PCP
Description
PMS2 Monoclonal antibody specifically detects PMS2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
PMS2 | |
Monoclonal | |
PerCP | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
OTI2G5 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
DNA mismatch repair protein PMS2, EC 3.1, H_DJ0042M02.9, HNPCC4postmeiotic segregation increased (S. cerevisiae) 2, mismatch repair endonuclease PMS2, PMS1 protein homolog 2, PMS2 postmeiotic segregation increased 2 (S. cerevisiae), PMSL2PMS2CL | |
Full length human recombinant protein of human PMS2(NP_000526) produced in HEK293T cell. | |
0.1 mL | |
Cancer, Cell Cycle and Replication, DNA Repair, Mismatch Repair | |
5395 | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction