Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PMS2 Antibody (PMS2/4373R) - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308570
Description
PMS2 Monoclonal specifically detects PMS2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Protein Array.Specifications
PMS2 | |
Monoclonal | |
1 mg/mL | |
Western Blot : 1-2 ug/mL, Flow Cytometry : 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence : 1-2 ug/mL, Immunohistochemistry-Paraffin, Protein Array | |
DNA mismatch repair protein PMS2, EC 3.1, H_DJ0042M02.9, HNPCC4postmeiotic segregation increased (S. cerevisiae) 2, mismatch repair endonuclease PMS2, PMS1 protein homolog 2, PMS2 postmeiotic segregation increased 2 (S. cerevisiae), PMSL2PMS2CL | |
A synthetic peptide corresponding to residues within aa1-100 of human PMS2 (exact sequence is proprietary) | |
100 μg | |
Cancer, Cell Cycle and Replication, DNA Repair, Mismatch Repair | |
5395 | |
Store at -20 to -80C. Avoid freeze-thaw cycles. | |
IgG κ |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Peptide Array | |
PMS2/4373R | |
Unconjugated | |
10mM PBS | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction