Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PMS2 Monoclonal Antibody (6G12E1)
GREENER_CHOICE

Catalog No. FERMA548576
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA548576 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA548576 Supplier Invitrogen™ Supplier No. MA548576
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PMS2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow, ELISA

PMS1 protein homologue 2 (PMS2) is a DNA mismatch repair protein. Defects in PMS2 are a cause of hereditary non-polyposis cancer (HNPCC). Human PMS2 is an amino acid protein of 862 amino acids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PMS2
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 6G12E1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene PMS2
Gene Accession No. P54278, P54279
Gene Alias AW555130; DNA mismatch repair protein PMS2; H_DJ0042M02.9; HNPCC4; mismatch repair endonuclease PMS2; MLH4; PMS1 homolog 2, mismatch repair protein; PMS1 homolog 2, mismatch repair system component; PMS1 protein homolog 2; PMS2; PMS2 postmeiotic segregation increased 2; PMS2 postmeiotic segregation increased 2 (S. cerevisiae); PMS2CL; PMSL2; postmeiotic segregation increased 2; postmeiotic segregation increased 2 (S. cerevisiae); postmeiotic segregation increased 2 nirs variant 6
Gene Symbols PMS2
Host Species Mouse
Immunogen Purified recombinant fragment of human PMS2 (AA: (431-580)) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18861, 5395
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.