Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PMS2 Mouse anti-Human, Clone: OTI2G5, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA506658S

Description
Mismatch DNA repair in bacteria is carried out by the MutL, MutH, and MutS proteins. The MutS protein initially binds to mismatched DNA. This is followed by binding of the MutH endonuclease and MutL to form a complex that carries out excision repair. The hMSH2 gene specifies a MutS homologue and hMLH1, hPMS1, and hPMS2 encode MutL homologs. Mutations in these genes are associated with hereditary nonpolyposis colon cancer (HNPCC), one of the most common hereditary diseases in man. As with the bacterial system, HNPCC is characterized by frequent microsatellite mutations that arise by somatic mutation due to a replication error (RER+) phenotype. Both hPMS1 and hPMS2 are mutated in the germline of HNPCC patients. Although the exact function of MutL and its homologs has yet to be determined, it is known that a complex of PMS2 and MLH1 (MutLa) from HeLa cells can complement a deficiency of MLH1 in hypermutable H6 colorectal tumor cells.Specifications
PMS2 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P54278 | |
PMS2 | |
Full length human recombit protein of human PMS2 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI2G5 | |
Unconjugated | |
PMS2 | |
HNPCC4, MLH4, PMS2CL, PMSL2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
5395 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction