Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PMS2 Mouse anti-Human, Clone: OTI2G5, liquid, TrueMAB™
SDP

Catalog No. 501676713
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-6713 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-6713 Supplier OriGene Supplier No. TA506658
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mismatch DNA repair in bacteria is carried out by the MutL, MutH, and MutS proteins. The MutS protein initially binds to mismatched DNA. This is followed by binding of the MutH endonuclease and MutL to form a complex that carries out excision repair. The hMSH2 gene specifies a MutS homologue and hMLH1, hPMS1, and hPMS2 encode MutL homologs. Mutations in these genes are associated with hereditary nonpolyposis colon cancer (HNPCC), one of the most common hereditary diseases in man. As with the bacterial system, HNPCC is characterized by frequent microsatellite mutations that arise by somatic mutation due to a replication error (RER+) phenotype. Both hPMS1 and hPMS2 are mutated in the germline of HNPCC patients. Although the exact function of MutL and its homologs has yet to be determined, it is known that a complex of PMS2 and MLH1 (MutLa) from HeLa cells can complement a deficiency of MLH1 in hypermutable H6 colorectal tumor cells.

Specifications

Antigen PMS2
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2G5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PMS2
Gene Accession No. P54278
Gene Alias HNPCC4, MLH4, PMS2CL, PMSL2
Gene Symbols PMS2
Host Species Mouse
Immunogen Full length human recombit protein of human PMS2 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5395
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.