Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PMS2 Mouse anti-Human, Mouse, Rat, Clone: 1G4E6, Proteintech
SDP

Catalog No. 501736512 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6512 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6512 Supplier Proteintech Group Inc Supplier No. 660751IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PMS1 protein homologue 2 (PMS2) is a DNA mismatch repair protein. Defects in PMS2 are a cause of hereditary non-polyposis cancer (HNPCC). Human PMS2 is an amino acid protein of 862 amino acids.

Specifications

Antigen PMS2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1G4E6
Concentration 1.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PMS2
Gene Accession No. P54278, P54279
Gene Alias HNPCC4, PMS1 protein homolog 2, PMS2, PMS2CL, PMSL2
Gene Symbols Pms2
Host Species Mouse
Immunogen PMS2 Fusion Protein Ag12661
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18861, 288479, 5395
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.