Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PMS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589357
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589357 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589357 Supplier Invitrogen™ Supplier No. PA589357
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ADLEKPMVEK QDQSPSLRTG EEKKDVSISR LREAFSLRHT TENKPHSPKT PEPRRSPLGQ KRGMLSSSTS GAISDKGVLR PQKEAVSSSH GPSDPTDRAE VEKDSGHGST SVDSEGFSIP DTGSHCSSEY AASSPGDRGS QEHVDSQEKA PKTDDSFSDV DCHSNQEDTG CKFRVLPQPT NLATPNTKRF KKEEILSSSD ICQKLVNTQD MSASQVDVAV KINKKVVPLD FSMSSLAKRI KQLHHEAQQS EGEQNYRKFR AKICPGENQA AEDELRKEIS K; Positive Samples: Jurkat, A-431, HeLa; Cellular Location: Nucleus.

PMS1 protein homologue 2 (PMS2) is a DNA mismatch repair protein. Defects in PMS2 are a cause of hereditary non-polyposis cancer (HNPCC). Human PMS2 is an amino acid protein of 862 amino acids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PMS2
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.36 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PMS2
Gene Accession No. P54278, P54279
Gene Alias AW555130; DNA mismatch repair protein PMS2; H_DJ0042M02.9; HNPCC4; mismatch repair endonuclease PMS2; MLH4; PMS1 homolog 2, mismatch repair protein; PMS1 homolog 2, mismatch repair system component; PMS1 protein homolog 2; PMS2; PMS2 postmeiotic segregation increased 2; PMS2 postmeiotic segregation increased 2 (S. cerevisiae); PMS2CL; PMSL2; postmeiotic segregation increased 2; postmeiotic segregation increased 2 (S. cerevisiae); postmeiotic segregation increased 2 nirs variant 6
Gene Symbols PMS2
Host Species Rabbit
Immunogen Recombinant protein (or fragment)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18861, 5395
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.