Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PMSCL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559749
Description
Immunogen sequence: CVVAGEKVWQ IRVDLHLLNH DGNIIDAASI AAIVALCHFR RPDVSVQGDE VTLYTPEERD PVPLSIHHMP ICVSFA Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
This gene encodes a component of the human exosome, a exoribonuclease complex which processes and degrades RNA in the nucleus and cytoplasm. This component may play a role in mRNA degradation and the polymyositis/scleroderma autoantigen complex. Alternative splicing results in multiple transcript variants.
Specifications
PMSCL1 | |
Polyclonal | |
Unconjugated | |
EXOSC9 | |
autoantigen PM/Scl 1; EXOSC9; Exosome complex component RRP45; exosome complex exonuclease RRP45; exosome component 9; exosome component; PM/Scl - 75; p5; p6; P75 polymyositis-scleroderma overlap syndrome associated autoantigen; P75 polymyositis-scleroderma overlap syndrome-associated autoantigen; PM/Scl-75; PMSCL autoantigen, 75kD; PMSCL1; Polymyositis/scleroderma autoantigen 1; polymyositis/scleroderma autoantigen 1, 75kDa; polymyositis/scleroderma autoantigen 75 kDa; RRP45; Rrp45p | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5393 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q06265 | |
EXOSC9 | |
Recombinant protein corresponding to Human PMSCL1. Recombinant protein control fragment (Product #RP-96639). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction