Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PMSCL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572959
Description
PMSCL1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IP, ChIP
This gene encodes a component of the human exosome, a exoribonuclease complex which processes and degrades RNA in the nucleus and cytoplasm. This component may play a role in mRNA degradation and the polymyositis/scleroderma autoantigen complex. Alternative splicing results in multiple transcript variants.
Specifications
PMSCL1 | |
Polyclonal | |
Unconjugated | |
EXOSC9 | |
autoantigen PM/Scl 1; EXOSC9; Exosome complex component RRP45; exosome complex exonuclease RRP45; exosome component 9; exosome component; PM/Scl - 75; p5; p6; P75 polymyositis-scleroderma overlap syndrome associated autoantigen; P75 polymyositis-scleroderma overlap syndrome-associated autoantigen; PM/Scl-75; PMSCL autoantigen, 75kD; PMSCL1; Polymyositis/scleroderma autoantigen 1; polymyositis/scleroderma autoantigen 1, 75kDa; polymyositis/scleroderma autoantigen 75 kDa; RRP45; Rrp45p | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
50911, 5393, 619530 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ChIP Assay, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
0.6 mg/mL | |
PBS with 30% glycerol and 0.05% sodium azide | |
Q06265, Q9JHI7 | |
EXOSC9 | |
A genomic peptide made to an internal region of human Exosome Component 9 (within residues 250-439). [Swiss-Prot Q06265]. | |
100 μL | |
Primary | |
Human, Mouse, Chicken | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction