Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PMVK Monoclonal Antibody (OTI2D1)
GREENER_CHOICE

Catalog No. PIMA525674
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525674 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525674 Supplier Invitrogen™ Supplier No. MA525674
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PMVK Monoclonal Antibody for Western Blot

PMVK is a peroxisomal enzyme that catalyzes the conversion of mevalonate 5-phosphate into mevalonate 5-diphosphate as the fifth reaction of the cholesterol biosynthetic pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PMVK
Applications Western Blot
Classification Monoclonal
Clone OTI2D1
Concentration 1.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Pmvk
Gene Accession No. Q15126, Q9D1G2
Gene Alias 1110011E12Rik; 2900002L22Rik; AV001327; hPMK; HUMPMKI; Phosphomevalonate kinase; PMK; PMKA; PMKASE; PMKI; Pmvk; POROK1; testis tissue sperm-binding protein Li 95mP
Gene Symbols Pmvk
Host Species Mouse
Immunogen Full length human recombinant protein of PMVK produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10654, 68603
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.