Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PMVK Monoclonal Antibody (OTI2D1), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA503448S


Description
PMVK Monoclonal antibody specifically detects PMVK in Human samples. It is validated for Western BlotSpecifications
PMVK | |
Monoclonal | |
1.35 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q15126 | |
Mouse | |
Affinity chromatography | |
RUO | |
10654 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2b |
Western Blot | |
OTI2D1 | |
Unconjugated | |
Pmvk | |
1110011E12Rik; 2900002L22Rik; AV001327; hPMK; HUMPMKI; Phosphomevalonate kinase; PMK; PMKA; PMKASE; PMKI; Pmvk; POROK1; testis tissue sperm-binding protein Li 95mP | |
Full length human recombinant protein of human PMVK produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction