Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PMVK/phosphomevalonate kinase Antibody (014), mFluor Violet 610 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290297MFV610
Description
PMVK/phosphomevalonate kinase Monoclonal antibody specifically detects PMVK/phosphomevalonate kinase in Human samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
PMVK/phosphomevalonate kinase | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunoprecipitation | |
014 | |
Western Blot, ELISA, Immunoprecipitation | |
hPMK, HUMPMKI, phosphomevalonate kinase, PMKA, PMKase, PMKEC 2.7.4.2, PMKI | |
Rabbit immunized with purified, recombinant Human PMVK/phosphomevalonate kinase (Uniprot#: Q15126, Met1-Leu192) | |
0.1 mL | |
Cancer, metabolism | |
10654 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction