Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PMVK/phosphomevalonate kinase Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | PMVK/phosphomevalonate kinase |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
PMVK/phosphomevalonate kinase Polyclonal specifically detects PMVK/phosphomevalonate kinase in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PMVK/phosphomevalonate kinase | |
Polyclonal | |
Rabbit | |
Cancer, metabolism | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
10654 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:PRLVLLFSGKRKSGKDFVTEALQSRLGADVCAVLRLSGPLKEQYAQEHGLNFQRLLDTSTYKEAFRKDMIRWGEEKRQADPGFFCRKIVEG | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
hPMK, HUMPMKI, phosphomevalonate kinase, PMKA, PMKase, PMKEC 2.7.4.2, PMKI | |
PMVK | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title