Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PMVK/phosphomevalonate kinase Antibody (OTI2F9), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273484H
Description
PMVK/phosphomevalonate kinase Monoclonal specifically detects PMVK/phosphomevalonate kinase in Human, Rat samples. It is validated for Western Blot.Specifications
PMVK/phosphomevalonate kinase | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
PMVK | |
Full length recombinant protein of human PMVK/phosphomevalonate kinase (NP_006547) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Rat | |
IgG2a |
Western Blot | |
OTI2F9 | |
Western Blot | |
hPMK, HUMPMKI, phosphomevalonate kinase, PMKA, PMKase, PMKEC 2.7.4.2, PMKI | |
Mouse | |
Affinity Purified | |
Cancer, metabolism | |
10654.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction