Learn More
PMVK Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305576AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine
PMVK is a peroxisomal enzyme that catalyzes the conversion of mevalonate 5-phosphate into mevalonate 5-diphosphate as the fifth reaction of the cholesterol biosynthetic pathway.Specifications
PMVK | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q15126, Q9D1G2 | |
PMVK | |
Between 142 and 192 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot | |
Unconjugated | |
PMVK | |
Phosphomevalonate kinase; PMKase; PMVK; PMKI; | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10654, 68603 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.