Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ PMXCEL™ Safety Eyewear

Sporty, lightweight design eliminates the bulkiness of a goggle

$33.40

Specifications

Certifications/Compliance ANSI Z87.1
Includes Strap
Weight (Metric) 32 g
Lens Material Polycarbonate
Frame Material Polycarbonate
View More Specs

Products 3
Catalog Number Mfr. No. Lens Tint dimLensCoating Price Quantity & Availability  
Catalog Number Mfr. No. Lens Tint dimLensCoating Price Quantity & Availability  
19-155-820
View Documents
Pyramex™
SB7321DT
Gray
Each for $33.40
Only null left
Add to Cart
 
19-155-818
View Documents
Pyramex™
SB7310DT
Clear N/A N/A N/A
19-155-819
View Documents
Pyramex™
SB7320DT
Gray N/A N/A N/A
Description

Description

  • Flame resistant inner foam padding
  • Durable frame with straight back temples and detachable strap
  • Anti-fog lenses
  • Scratch resistant polycarbonate lens provides 99% UVA/B/C protection
  • Specifications

    Specifications

    ANSI Z87.1
    32 g
    Polycarbonate
    Strap
    Polycarbonate
    Videos
    SDS
    Documents

    Documents

    Product Certifications
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.