Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PNAS4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565589
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565589 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565589 Supplier Thermo Scientific Supplier No. PA565589
Add to Cart
Add to Cart

PNAS4 Polyclonal Antibody

PNAS4 Polyclonal Antibody for ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen PNAS4
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DESI2
Gene Accession No. Q9BSY9
Gene Alias 5830417C01Rik; C1orf121; cb238; CGI-146; DESI; DESI1; desi2; DeSI-2; desumoylating isopeptidase 1; desumoylating isopeptidase 2; Deubiquitinase DESI2; Fam152a; family with sequence similarity 152, member A; fi12d12; hypothetical protein LOC445138; pnas4; PNAS-4; PPPDE peptidase domain containing 1; PPPDE peptidase domain-containing protein 1; Pppde1; Protein FAM152A; sb:cb238; si:ch211-132e22.3; wu:fi12d12; zgc:100860
Gene Symbols DESI2
Host Species Rabbit
Immunogen Recombinant Human PNAS4. Recombinant protein control fragment (Product #RP-99772).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51029
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.