Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PNAS4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106335
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106335 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106335 Supplier Invitrogen™ Supplier No. PA5106335
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PPPDE1/PNAS4.

PNAS4 is a highly conserved protein that shares high homology from plants to animals. It was initially identified as a putative apoptosis-related protein in the human acute promyelocytic leukemia cell line NB4. PNAS4 is activated during the early response to DNA damage and can inhibit proliferation via apoptosis when overexpressed in some tumor cells such as U2OS, SKOV3, and A549. PNAS4 inhibits tumor cell proliferation through the following mechanisms: (i) overexpression of PNAS4 causes S phase arrest by regulating the expression of cell cycle-related proteins and (ii) PNAS4 induces apoptosis through the mitochondrial apoptosis pathway. Recent evidence has shown that PNAS4 may be involved in the genesis of some cancers and could be a potential candidate for lung cancer therapy alone or in combination with gemcitabine. At least two isoforms of PNAS4 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PNAS4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DESI2
Gene Accession No. Q5XIT6, Q9BSY9, Q9D291
Gene Alias 5830417C01Rik; C1orf121; cb238; CGI-146; DESI; DESI1; desi2; DeSI-2; desumoylating isopeptidase 1; desumoylating isopeptidase 2; Deubiquitinase DESI2; Fam152a; family with sequence similarity 152, member A; fi12d12; hypothetical protein LOC445138; pnas4; PNAS-4; PPPDE peptidase domain containing 1; PPPDE peptidase domain-containing protein 1; Pppde1; Protein FAM152A; sb:cb238; si:ch211-132e22.3; wu:fi12d12; zgc:100860
Gene Symbols DESI2
Host Species Rabbit
Immunogen A synthesized peptide derived from human DESI2(Accession Q9BSY9), corresponding to amino acid residues T21-E71.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 289277, 51029, 78825
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.