Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PNCK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552437
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552437 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552437 Supplier Invitrogen™ Supplier No. PA552437
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MLLLKKHTED ISSVYEIRER LGSGAFSEVV LAQERGSAHL VALKCIPKKA LRGKEALVEN EIAVLRRISH PN Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

PNCK is a member of the calcium/calmodulin-dependent protein kinase family of protein serine/threonine kinases (Gardner et al., 2000 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PNCK
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PNCK
Gene Accession No. Q6P2M8
Gene Alias BSTK3; calcium/calmodulin-dependent protein kinase 1 beta; calcium/calmodulin-dependent protein kinase 1, beta; calcium/calmodulin-dependent protein kinase type 1B; CaM kinase I beta; caM kinase IB; CaMK1b; caM-KI beta; caMKI-beta; CaMKIbeta2; caMKlb1; CaMKlb2; Pnck; pregnancy up-regulated nonubiquitous CaM kinase; pregnancy up-regulated non-ubiquitously expressed CaM kinase; pregnancy upregulated non-ubiquitously expressed CaM kinase; pregnancy up-regulated non-ubiquitously-expressed CaM kinase; Pregnancy up-regulated non-ubiquitously-expressed CaM kinase homolog; Punc
Gene Symbols PNCK
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PNCK. Recombinant protein control fragment (Product #RP-89783).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 139728
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.