Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PNCK Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598524
Description
PNCK Polyclonal Antibody for Western Blot, IHC (P), ELISA
PNCK is a member of the calcium/calmodulin-dependent protein kinase family of protein serine/threonine kinases (Gardner et al., 2000 ).
Specifications
PNCK | |
Polyclonal | |
Unconjugated | |
PNCK | |
BSTK3; calcium/calmodulin-dependent protein kinase 1 beta; calcium/calmodulin-dependent protein kinase 1, beta; calcium/calmodulin-dependent protein kinase type 1B; CaM kinase I beta; caM kinase IB; CaMK1b; caM-KI beta; caMKI-beta; CaMKIbeta2; caMKlb1; CaMKlb2; Pnck; pregnancy up-regulated nonubiquitous CaM kinase; pregnancy up-regulated non-ubiquitously expressed CaM kinase; pregnancy upregulated non-ubiquitously expressed CaM kinase; pregnancy up-regulated non-ubiquitously-expressed CaM kinase; Pregnancy up-regulated non-ubiquitously-expressed CaM kinase homolog; Punc | |
Rabbit | |
Protein G | |
RUO | |
139728, 29660, 93843 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
3.13 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
O70150, Q6P2M8, Q9QYK9 | |
PNCK | |
Recombinant Human Calcium/calmodulin-dependent protein kinase type 1B protein (1-115AA). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction