Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Pneumolysin Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144564
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144564 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144564 Supplier Invitrogen™ Supplier No. PA5144564
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Pneumolysin Polyclonal Antibody for Western Blot, ELISA

Pneumolysin (pneumococcal hemolysin) is an intracellular protein produced by all strains of S. pneumoniae and is a cytoplasmic cholesterol-dependent cytolysin forming large pores in cholesterol-containing membranes and are therefore cytotoxic to mammalian cells. The molecular weight of Pneumolysin is approximately 53 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pneumolysin
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene ply
Gene Accession No. Q04IN8
Gene Alias ply; Pneumolysin (Thiol-activated cytolysin)
Gene Symbols ply
Host Species Rabbit
Immunogen Recombinant Streptococcus pneumoniae serotype 2 ply protein (2-471aa).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66806991
Target Species Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.