Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PNK1/PNKP Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985481
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5481 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5481 Supplier Bioss Supplier No. BS3542R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-human Polynucleotide Kinase Mouse Monoclonal Antibody recognizes polynucleotide kinase (PNK), a 57 kDa nuclear enzyme implicated in DNA strand repair processes that occur during DNA replication and recombination, or in response to DNA damaging agents. PNK targets the DNA 5′-hydroxy and 3′-phosphate termini for phosphorylation and dephosphorylation events. During strand breakage in human cells, normal PNK associates with other nuclear proteins to prepare the DNA termini for the enzymatic actions of DNA polymerase-beta and DNA ligase III. PNK is a key functional component in maintaining the genetic stability of human cells.

Specifications

Antigen PNK1/PNKP
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Pnkp
Gene Accession No. Q96T60
Gene Alias 1810009G08Rik; 2′(3′)-polynucleotidase; AOA4; Bifunctional polynucleotide phosphatase/kinase; DNA 5′-kinase/3′-phosphatase; EIEE10; Homo sapiens polynucleotide kinase 3′-phosphatase (PNKP); MCSZ; phosphatase/kinase; PKNP; PNK; PNKP; Polynucleotide 3′-phosphatase; Polynucleotide 5′-hydroxyl-kinase; polynucleotide kinase 3′- phosphatase; polynucleotide kinase 3′-phosphatase; Polynucleotide kinase-3′-phosphatase
Gene Symbols Pnkp
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PNK1/PNKP.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11284
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.